Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zp3b
Entrez Gene ID
64692
Gene Name
zona pellucida glycoprotein 3b
Gene Aliases
ZP3, ZPC, fa55e05, wu:fa55e05, zfZPC, zpc6
Chromosome Location
Chr.2: 6253291 - 6258794 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131696.1 NP_571771.1 1 - 2 506 114
GenBank mRNA BC067692.1 - 2 - 3 456 114
BC151892.1 - 1 - 2 508 114
U55863.3 - 1 - 2 506 114

View Full Product Details