Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
jag1b
Entrez Gene ID
140423
Gene Name
jagged canonical Notch ligand 1b
Gene Aliases
jag3, kmi, puc, unp1069
Chromosome Location
Chr.13: 35339182 - 35390095 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131863.2 NP_571938.2 13 - 14 1971 59
GenBank mRNA AF229451.1 - 13 - 14 1933 59
AY221106.1 - 13 - 14 1972 59

View Full Product Details