Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
jag1b
Entrez Gene ID
140423
Gene Name
jagged canonical Notch ligand 1b
Gene Aliases
jag3, kmi, puc, unp1069
Chromosome Location
Chr.13: 35339182 - 35390095 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131863.2 NP_571938.2 6 - 7 1136 78
GenBank mRNA AF229451.1 - 6 - 7 1098 78
AY221106.1 - 6 - 7 1137 78

View Full Product Details