Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp2ad2
Entrez Gene ID
259306
Gene Name
cytochrome P450, family 2, subfamily AD, polypeptide 2
Gene Aliases
cyp2j28, cyp2ja, fb62g08, wu:fb62g08
Chromosome Location
Chr.20: 25534672 - 25542171 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152954.1 NP_694486.1 2 - 3 386 69
GenBank mRNA AF248042.2 - 2 - 3 385 69
BC045948.1 - 2 - 3 403 69
BC165354.1 - 2 - 3 381 69

View Full Product Details