Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp2ad2
Entrez Gene ID
259306
Gene Name
cytochrome P450, family 2, subfamily AD, polypeptide 2
Gene Aliases
cyp2j28, cyp2ja, fb62g08, wu:fb62g08
Chromosome Location
Chr.20: 25534672 - 25542171 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152954.1 NP_694486.1 7 - 8 1198 83
GenBank mRNA AF248042.2 - 7 - 8 1197 83

View Full Product Details