Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
trpn1
Entrez Gene ID
368273
Gene Name
transient receptor potential cation channel, subfamily N, member 1
Gene Aliases
nompC
Chromosome Location
Chr.19: 1435878 - 1479738 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183349.1 NP_899192.1 15 - 15 1980 81
GenBank mRNA AY313897.1 - 15 - 15 1980 81

View Full Product Details