Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
trpn1
Entrez Gene ID
368273
Gene Name
transient receptor potential cation channel, subfamily N, member 1
Gene Aliases
nompC
Chromosome Location
Chr.19: 1435878 - 1479738 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183349.1 NP_899192.1 11 - 12 1406 112
GenBank mRNA AY313897.1 - 11 - 12 1406 112

View Full Product Details