Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fem1c
Entrez Gene ID
378965
Gene Name
fem-1 homolog c
Gene Aliases
-
Chromosome Location
Chr.8: 423393 - 425258 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198145.2 NP_937788.2 - 824 79
GenBank mRNA AY249190.1 - - 545 79
BC056774.1 - - 824 79

View Full Product Details