Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
polr2g
Entrez Gene ID
324088
Gene Name
RNA polymerase II subunit G
Gene Aliases
polr2gl, wu:fc18d05, zgc:55297
Chromosome Location
Chr.14: 26715644 - 26719053 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199669.1 NP_955963.1 2 - 3 225 125
GenBank mRNA BC045299.1 - 2 - 3 225 125
BC164883.1 - 1 - 2 147 125

View Full Product Details