Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
cdo1
Entrez Gene ID
393714
Gene Name
cysteine dioxygenase, type I
Gene Aliases
CDO-I, cdo, cdo-1, zgc:73181, zgc:77945
Chromosome Location
Chr.Unknown: 4031 - 7255 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200741.1 NP_957035.2 - 429 88
GenBank mRNA BC059531.1 - - 429 88
BC067344.1 - - 88

View Full Product Details