Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sult1st4
Entrez Gene ID
402915
Gene Name
sulfotransferase family 1, cytosolic sulfotransferase 4
Gene Aliases
zgc:76922
Chromosome Location
Chr.8: 53171006 - 53180060 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205620.2 NP_991183.1 3 - 4 341 92
XM_021478222.1 XP_021333897.1 3 - 4 511 92
XM_021478223.1 XP_021333898.1 3 - 4 820 92
XM_021478224.1 XP_021333899.1 3 - 4 497 92
GenBank mRNA AY196986.1 - 3 - 4 298 92
BC066584.1 - 3 - 4 341 92

View Full Product Details