Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
klhl21
Entrez Gene ID
404613
Gene Name
kelch-like family member 21
Gene Aliases
zgc:76933
Chromosome Location
Chr.23: 24926353 - 24939148 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320407.1 NP_001307336.1 5 - 6 1729 93
XM_005162160.4 XP_005162217.1 6 - 7 1884 93
XM_021469772.1 XP_021325447.1 4 - 5 2235 93
GenBank mRNA BC066537.1 - 3 - 4 1734 93
BC155818.1 - 5 - 6 1729 93

View Full Product Details