Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dip2ba
Entrez Gene ID
405899
Gene Name
disco-interacting protein 2 homolog Ba
Gene Aliases
dip2b, zgc:77348
Chromosome Location
Chr.23: 27358722 - 27442571 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212963.1 NP_998128.1 12 - 13 1685 71
XM_009296840.3 XP_009295115.1 13 - 14 1729 71
XM_009296841.3 XP_009295116.1 12 - 13 1723 71
XM_009296842.3 XP_009295117.1 13 - 14 1729 71
GenBank mRNA BC068015.1 - 12 - 13 1685 71

View Full Product Details