Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pias4a
Entrez Gene ID
406712
Gene Name
protein inhibitor of activated STAT, 4a
Gene Aliases
pias4, piasy, wu:fc54c11, wu:fi18c10, wu:fi20e09, wu:fk93f11, zgc:66410
Chromosome Location
Chr.22: 20406858 - 20419670 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213403.2 NP_998568.2 9 - 10 1116 94
GenBank mRNA BC057528.1 - 9 - 10 1106 94
BC070017.1 - 9 - 10 1116 94
JF759916.1 - 9 - 10 1135 94

View Full Product Details