Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gins3
Entrez Gene ID
442931
Gene Name
GINS complex subunit 3
Gene Aliases
flj13912, zgc:100834
Chromosome Location
Chr.25: 13399967 - 13403655 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002874.1 NP_001002874.1 2 - 3 486 84
GenBank mRNA AY648732.1 - 2 - 3 486 84
BC078185.1 - 2 - 3 492 84

View Full Product Details