Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pomp
Entrez Gene ID
445030
Gene Name
proteasome maturation protein
Gene Aliases
fa13f09, wu:fa13f09, zgc:92775
Chromosome Location
Chr.21: 40712883 - 40717636 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003424.2 NP_001003424.1 5 - 6 390 85
GenBank mRNA BC076250.1 - 5 - 6 392 85

View Full Product Details