Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
xgb
Entrez Gene ID
497122
Gene Name
x globin
Gene Aliases
-
Chromosome Location
Chr.17: 1664625 - 1703259 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012261.2 NP_001012261.2 5 - 6 640 67
XM_021467154.1 XP_021322829.1 4 - 5 690 67
GenBank mRNA BC133862.1 - 5 - 6 640 67

View Full Product Details