Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pygma
Entrez Gene ID
553655
Gene Name
phosphorylase, glycogen, muscle A
Gene Aliases
im:7150327, zgc:110706
Chromosome Location
Chr.21: 28445052 - 28472235 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020628.1 NP_001018464.1 13 - 14 1682 62
GenBank mRNA BC095379.1 - 13 - 14 1682 62

View Full Product Details