Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tvp23b
Entrez Gene ID
554134
Gene Name
trans-golgi network vesicle protein 23 homolog B (S. cerevisiae)
Gene Aliases
fam18b, zgc:110017
Chromosome Location
Chr.12: 46683216 - 46696041 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024428.1 NP_001019599.1 2 - 3 119 99
GenBank mRNA BC095132.1 - 2 - 3 119 99
BC165009.1 - 1 - 2 107 99

View Full Product Details