Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mcm6l
Entrez Gene ID
564982
Gene Name
MCM6 minichromosome maintenance deficient 6, like
Gene Aliases
MCM6m, Mcm6, zgc:112244
Chromosome Location
Chr.2: 299059 - 323178 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025533.1 NP_001020704.1 7 - 8 1114 72
GenBank mRNA AB973821.1 - 7 - 8 1061 72
BC096872.1 - 7 - 8 1114 72

View Full Product Details