Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
clvs2
Entrez Gene ID
566769
Gene Name
clavesin 2
Gene Aliases
si:ch211-199i15.4
Chromosome Location
Chr.20: 40269902 - 40319890 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025545.2 NP_001020716.1 1 - 2 813 92
GenBank mRNA BC116475.1 - 1 - 2 813 92

View Full Product Details