Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bco2b
Entrez Gene ID
678554
Gene Name
beta-carotene oxygenase 2b
Gene Aliases
bcdo2, wu:fk75b02, zgc:136769
Chromosome Location
Chr.15: 21673454 - 21678634 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040312.1 NP_001035402.1 7 - 8 1005 91
GenBank mRNA BC115259.1 - 7 - 8 1005 91

View Full Product Details