Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srrd
Entrez Gene ID
751717
Gene Name
SRR1 domain containing
Gene Aliases
zgc:153763
Chromosome Location
Chr.8: 4434018 - 4444568 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045427.1 NP_001038892.1 3 - 4 519 112
XM_005172446.4 XP_005172503.1 3 - 4 112
XM_009303729.3 XP_009302004.1 3 - 4 112
GenBank mRNA BC122414.1 - 3 - 4 519 112

View Full Product Details