Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
jmjd4
Entrez Gene ID
767690
Gene Name
jumonji domain containing 4
Gene Aliases
CH1073-162B20.2, zgc:153974
Chromosome Location
Chr.2: 3403264 - 3414068 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076628.1 NP_001070096.1 1 - 2 602 76
GenBank mRNA BC124503.1 - 1 - 2 602 76

View Full Product Details