Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hnrnpd
Entrez Gene ID
560522
Gene Name
heterogeneous nuclear ribonucleoprotein D
Gene Aliases
wu:fa28b06, zgc:162951
Chromosome Location
Chr.10: 54967 - 71417 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110460.1 NP_001103930.1 3 - 4 654 145
XM_005155544.3 XP_005155601.1 3 - 4 677 145
XM_005155545.3 XP_005155602.1 3 - 4 677 145
XM_005155546.3 XP_005155603.1 3 - 4 677 145
XM_005155547.3 XP_005155604.1 3 - 4 677 145
XM_021479184.1 XP_021334859.1 3 - 4 677 145
GenBank mRNA BC139674.1 - 3 - 4 654 145

View Full Product Details