Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
plekho1a
Entrez Gene ID
563283
Gene Name
pleckstrin homology domain containing, family O member 1a
Gene Aliases
ckip-1, plekho1, si:dkey-200h23.2
Chromosome Location
Chr.19: 42367286 - 42391635 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110463.1 NP_001103933.1 4 - 5 526 68
GenBank mRNA BC134933.1 - 4 - 5 526 68

View Full Product Details