The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_131292.1 | NP_571367.1 | 26 - 27 | 1786 | 61 |
GenBank mRNA | BC163934.1 | - | 26 - 27 | 1778 | 61 |
DQ335127.1 | - | 26 - 27 | 1786 | 61 |