Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mpp5a
Entrez Gene ID
252845
Gene Name
membrane protein, palmitoylated 5a (MAGUK p55 subfamily member 5)
Gene Aliases
mpp5, nok
Chromosome Location
Chr.17: 34186605 - 34239519 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194363.1 NP_919344.1 14 - 15 2277 92
XM_009293174.3 XP_009291449.1 15 - 16 2379 92
GenBank mRNA AF510111.1 - 14 - 15 2277 92
BC163251.1 - 12 - 13 2011 92
BC163277.1 - 12 - 13 2011 92

View Full Product Details