Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rbm18
Entrez Gene ID
393766
Gene Name
RNA binding motif protein 18
Gene Aliases
zgc:73332
Chromosome Location
Chr.10: 133806 - 158367 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200793.1 NP_957087.1 2 - 3 273 110
XM_005155615.4 XP_005155672.1 2 - 3 274 110
XM_009305144.3 XP_009303419.1 2 - 3 274 110
GenBank mRNA BC059651.1 - 2 - 3 273 110

View Full Product Details