Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bhlhe22
Entrez Gene ID
393930
Gene Name
basic helix-loop-helix family, member e22
Gene Aliases
Beta3.1, bhlhb5, wu:fq25e11, zgc:64225
Chromosome Location
Chr.24: 24461558 - 24463346 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200955.1 NP_957249.2 1 - 1 849 83
GenBank mRNA AY581209.1 - 1 - 1 191 83
BC053312.1 - 1 - 1 849 83

View Full Product Details