Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
atf3
Entrez Gene ID
393939
Gene Name
activating transcription factor 3
Gene Aliases
zgc:55526
Chromosome Location
Chr.20: 37794678 - 37800737 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200964.1 NP_957258.1 2 - 3 343 64
GenBank mRNA BC045371.1 - 2 - 3 343 64

View Full Product Details