Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc33a1
Entrez Gene ID
394083
Gene Name
solute carrier family 33 member 1
Gene Aliases
zgc:63693
Chromosome Location
Chr.18: 34156598 - 34170941 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201108.1 NP_957402.1 2 - 3 891 126
XM_005159215.4 XP_005159272.1 2 - 3 996 126
XM_021467624.1 XP_021323299.1 2 - 3 904 126
XM_021467625.1 XP_021323300.1 2 - 3 847 126
GenBank mRNA BC055208.1 - 2 - 3 891 126

View Full Product Details