Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pdcd5
Entrez Gene ID
394152
Gene Name
programmed cell death 5
Gene Aliases
zgc:56315
Chromosome Location
Chr.18: 31089950 - 31105391 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201177.1 NP_957471.1 4 - 5 522 122
GenBank mRNA BC046067.1 - 4 - 5 522 122
BC071345.1 - 4 - 5 517 122

View Full Product Details