Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pvalb1
Entrez Gene ID
402805
Gene Name
parvalbumin 1
Gene Aliases
zgc:86939
Chromosome Location
Chr.3: 61193890 - 61205783 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205572.1 NP_991135.1 3 - 4 326 113
XM_005164288.3 XP_005164345.1 4 - 5 443 113
GenBank mRNA AF467914.1 - 3 - 4 326 113
BC071552.1 - 4 - 5 366 113

View Full Product Details