Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
glud1a
Entrez Gene ID
317737
Gene Name
glutamate dehydrogenase 1a
Gene Aliases
cb622, glud1, wu:fc33g09, wu:fc66a10, zgc:77186
Chromosome Location
Chr.13: 22679083 - 22699024 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212576.1 NP_997741.1 5 - 6 811 70
GenBank mRNA BC066370.1 - 5 - 6 811 70

View Full Product Details