Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ywhah
- Entrez Gene ID
-
406443
- Gene Name
- tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, eta polypeptide
- Gene Aliases
-
fb93e02, wu:fb51h04, wu:fb93e02, ywhag, zgc:63883
- Chromosome Location
-
Chr.10: 17166035 - 17170086 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_213164.2 |
NP_998329.1 |
1 - 2 |
403 |
84 |
GenBank mRNA |
BC053130.1 |
- |
1 - 2 |
403 |
84 |
View Full Product Details