Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
commd2
Entrez Gene ID
436694
Gene Name
COMM domain containing 2
Gene Aliases
wu:fb12e01, zgc:92665
Chromosome Location
Chr.22: 38037530 - 38045244 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002421.1 NP_001002421.1 4 - 5 511 92
GenBank mRNA BC076149.1 - 4 - 5 511 92

View Full Product Details