Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cbln1
Entrez Gene ID
436793
Gene Name
cerebellin 1 precursor
Gene Aliases
zgc:92807
Chromosome Location
Chr.18: 419600 - 420630 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002520.1 NP_001002520.1 2 - 3 600 90
XM_005159080.4 XP_005159137.1 2 - 3 977 90
GenBank mRNA BC076277.1 - 2 - 3 600 90

View Full Product Details