Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sec11a
Entrez Gene ID
436794
Gene Name
SEC11 homolog A, signal peptidase complex subunit
Gene Aliases
sec11, sec11l1, zgc:92805
Chromosome Location
Chr.18: 297103 - 299578 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002521.2 NP_001002521.1 2 - 3 279 135
GenBank mRNA BC076276.1 - 2 - 3 283 135

View Full Product Details