Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lygl1
Entrez Gene ID
436979
Gene Name
lysozyme g-like 1
Gene Aliases
LysG, zgc:92608
Chromosome Location
Chr.14: 20893031 - 20898504 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002706.1 NP_001002706.1 2 - 3 219 114
XM_005173116.3 XP_005173173.1 2 - 3 220 114
GenBank mRNA BC076099.1 - 2 - 3 219 114

View Full Product Details