Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc16a9b
Entrez Gene ID
445158
Gene Name
solute carrier family 16 member 9b
Gene Aliases
slc16a9, wu:fk66d11, zgc:101135
Chromosome Location
Chr.12: 7589190 - 7607112 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003552.1 NP_001003552.1 4 - 5 671 61
GenBank mRNA BC078337.1 - 4 - 5 671 61

View Full Product Details