Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
wdr54
Entrez Gene ID
445236
Gene Name
WD repeat domain 54
Gene Aliases
zgc:100930
Chromosome Location
Chr.5: 43072925 - 43083484 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003630.2 NP_001003630.2 8 - 9 1076 67
GenBank mRNA BC077087.1 - 8 - 9 1080 67

View Full Product Details