Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
akr1a1a
Entrez Gene ID
445326
Gene Name
aldo-keto reductase family 1, member A1a (aldehyde reductase)
Gene Aliases
akr1a1, zgc:100940
Chromosome Location
Chr.5: 290556 - 298112 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003783.2 NP_001003783.1 8 - 9 951 132
GenBank mRNA BC077140.1 - 8 - 9 955 132

View Full Product Details