Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
st3gal2
Entrez Gene ID
445562
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 2
Gene Aliases
si:ch211-223o1.7, siat5
Chromosome Location
Chr.18: 992 - 3219 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004012.1 NP_001004012.1 1 - 2 407 80
XM_009293548.3 XP_009291823.1 2 - 3 2172 80
XM_009293550.3 XP_009291825.1 2 - 3 1931 80
GenBank mRNA AJ705062.2 - 1 - 2 407 80
AJ783741.1 - 1 - 2 407 80
BC162919.1 - 1 - 2 456 80
BC162930.1 - 1 - 2 456 80

View Full Product Details