Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rbm8a
Entrez Gene ID
503767
Gene Name
RNA binding motif protein 8A
Gene Aliases
fb34f03, wu:fb34f03, zgc:110525
Chromosome Location
Chr.16: 45922224 - 45930077 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013345.1 NP_001013363.1 5 - 6 503 89
GenBank mRNA BC090261.1 - 5 - 6 503 89

View Full Product Details