Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
eif3k
Entrez Gene ID
550245
Gene Name
eukaryotic translation initiation factor 3, subunit K
Gene Aliases
zgc:110726
Chromosome Location
Chr.15: 47845368 - 47848890 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017583.1 NP_001017583.1 3 - 4 380 75
GenBank mRNA BC093002.1 - 3 - 4 380 75

View Full Product Details