Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wdr73
Entrez Gene ID
550339
Gene Name
WD repeat domain 73
Gene Aliases
zgc:112071
Chromosome Location
Chr.21: 21819878 - 21824289 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017646.1 NP_001017646.1 2 - 3 258 79
XM_009295306.3 XP_009293581.1 2 - 3 262 79
GenBank mRNA BC093184.1 - 2 - 3 258 79

View Full Product Details