Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxo16
Entrez Gene ID
550375
Gene Name
F-box protein 16
Gene Aliases
si:ch211-89p3.1, zgc:112464
Chromosome Location
Chr.20: 35247526 - 35254631 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017680.1 NP_001017680.1 5 - 6 676 97
GenBank mRNA BC093325.1 - 5 - 6 676 97
BC164389.1 - 4 - 5 497 97

Gene Symbol
LOC100150082
Entrez Gene ID
100150082
Gene Name
F-box protein 16
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128401.1 NP_001121873.1 5 - 6 792 97

View Full Product Details