Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
btbd6b
Entrez Gene ID
553533
Gene Name
BTB (POZ) domain containing 6b
Gene Aliases
btbd6, btbd6a, sb:cb776, si:dkey-11b8.2, zgc:152751
Chromosome Location
Chr.20: 20927827 - 20932782 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024815.2 NP_001019986.1 2 - 3 407 93
XM_005160494.4 XP_005160551.1 1 - 2 1079 93
GenBank mRNA BC095161.1 - 2 - 3 384 93
BC115089.1 - 2 - 3 407 93
BC124114.1 - 2 - 3 351 93

View Full Product Details