Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pip4p2
Entrez Gene ID
566642
Gene Name
phosphatidylinositol-4,5-bisphosphate 4-phosphatase 2
Gene Aliases
si:ch211-134n17.1, tmem55a
Chromosome Location
Chr.19: 362976 - 390844 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045112.3 NP_001038577.1 4 - 5 538 61

View Full Product Details